4-(Phenoxymethyl)benzoesäure

Lieferant: BIONET RESEARCH

10P-369S.500 10P-369S.1 10P-369S.5 10P-369S.10
BIOT10P-369S.500EA 0 EUR
BIOT10P-369S.500 BIOT10P-369S.1 BIOT10P-369S.5 BIOT10P-369S.10
4-(Phenoxymethyl)benzoesäure
4-(Phenoxymethyl)benzoic acid
Formel: C₁₄H₁₂O₃
Molecular Weight: 228.2433 g/mol
MDL: MFCD00623268
CAS-Nummer: 31719-76-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR