5-Brom-2-chlorisonicotinsäure

Lieferant: COMBI-BLOCKS

PY-7077-1G PY-7077-5G
COBBPY-7077-1GEA 33.5 EUR
COBBPY-7077-1G COBBPY-7077-5G
5-Brom-2-chlorisonicotinsäure
5-Brom-2-chlorisonicotinsäure
Formel: C₆H₃BrClNO₂
Molecular Weight: 236.4500 g/mol
MDL: MFCD07375114
CAS-Nummer: 886365-31-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR