3,5-Difluorisonicotinsäure

Lieferant: COMBI-BLOCKS

OT-8123-5G OT-8123-1G
COBBOT-8123-5GEA 190 EUR
COBBOT-8123-5G COBBOT-8123-1G
3,5-Difluorisonicotinsäure
3,5-Difluorisonicotinsäure
Formel: C₆H₃F₂NO₂
Molecular Weight: 159.0900 g/mol
MDL: MFCD04972829
CAS-Nummer: 903522-29-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR