Natriumheptafluorbutyrat

Lieferant: FLUOROCHEM

007276-5G 007276-25G
FLUO007276-5GEA 0 EUR
FLUO007276-5G FLUO007276-25G
Natriumheptafluorbutyrat
Natriumheptafluorbutyrat
Formel: C₄F₇NaO₂
Molecular Weight: 236,02 g/mol
MDL: MFCD00066168
CAS-Nummer: 2218-54-4
EINECS: 218-721-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR