tert-Butyl-2-bromisobutyrat ≥98%, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
tert-Butyl α-bromoisobutyrate, 2-Bromoisobutyric acid tert-butyl ester, tert-Butyl 2-methyl-2-propionate, tert-Butyl 2-bromo-2-methylpropanoate
17455-100ML
17455-500ML
SIAL17455-100MLEA
53.1
EUR
SIAL17455-100ML
SIAL17455-500ML
tert-Butyl-2-bromisobutyrat ≥98%, Sigma-Aldrich®
tert-Butyl-2-bromisobutyrat
tert-Butyl α-bromoisobutyrate was used as an initiator in the atom transfer radical polymerisation (ATRP) of 2-(acetoacetoxy)ethyl methacrylate (AEMA).
Formel:
C₆H₁₁BrO₂ Molecular Weight: 223,12 g/mol |
MDL:
MFCD00051908 CAS-Nummer: 23877-12-5 EINECS: 209-980-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...