3-Brom-1H-indazol ≥97%

Lieferant: Thermo Scientific
Gefahr

H37775.03 H37775.09
H37775.03EA 52.5 EUR
H37775.03 H37775.09
3-Brom-1H-indazol ≥97%
3-Bromo-1H-indazole
Formel: C₇H₅BrN₂
Molecular Weight: 197,04 g/mol
Schmelzpunkt: 148…149 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00159926
CAS-Nummer: 40598-94-5
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR