3-Chlor-2-fluor-5-(trifluormethyl)benzylbromid

Lieferant: FLUOROCHEM

005687-1G 005687-5G
FLUO005687-1GEA 0 EUR
FLUO005687-1G FLUO005687-5G
3-Chlor-2-fluor-5-(trifluormethyl)benzylbromid
3-Chlor-2-fluor-5-(trifluormethyl)benzylbromid
Formel: C₈H₄BrClF₄
MDL: MFCD01631513
CAS-Nummer: 261763-09-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR