6-Bromnicotinonitril
Lieferant: COMBI-BLOCKS
Synonyme:
2-Bromo-5-cyanopyridine
CA-4321-5G
CA-4321-25G
CA-4321-1G
COBBCA-4321-5GEA
53.1
EUR
COBBCA-4321-5G
COBBCA-4321-25G
COBBCA-4321-1G
6-Bromnicotinonitril
6-Bromnicotinonitril
Formel:
C₆H₃BrN₂ Molecular Weight: 183,01 g/mol |
MDL:
MFCD00234141 CAS-Nummer: 139585-70-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...