Methyl-4-tert-butylbenzoat ≥98%

Lieferant: Thermo Scientific

Synonyme: Methyl 4-(tert-butyl)benzoate

A18619.22 A18619.14
A18619.22EA 108 EUR
A18619.22 A18619.14
Methyl-4-tert-butylbenzoat ≥98%
Methyl-4-tert-butylbenzoat

Purity: 98+% CAS No.: 26537-19-9 Molecular Formula: C12H16O2 Formula Weight: 192.26 Flash Point: 110°(230°F) Refractive Index: 1.5100 Boiling Point: 122-124°/10mm MDL No.: MFCD00008835

Formel: C₁₂H₁₆O₂
Molecular Weight: 192,26 g/mol
Siedepunkt: 122…124 °C (10 mmHg)
Dichte: 0,995
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008835
CAS-Nummer: 26537-19-9
EINECS: 247-768-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR