Methyl-4-tert-butylbenzoat

Lieferant: CHEMPUR

Synonyme: Methyl 4-(tert-butyl)benzoate

S06946.250 S06946.1
CHMPS06946.250EA 0 EUR
CHMPS06946.250 MAYBS06946.1
Methyl-4-tert-butylbenzoat
Methyl-4-tert-butylbenzoat
Formel: C₁₂H₁₆O₂
Molecular Weight: 192,26 g/mol
MDL: MFCD00008835
CAS-Nummer: 26537-19-9
EINECS: 247-768-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR