Methyl-4-butylbenzoat
Lieferant: Thermo Scientific
TL00831DA
TL00831EA
TL00831EE
MAYBTL00831DAEA
0
EUR
MAYBTL00831DA
MAYBTL00831EA
MAYBTL00831EE
Methyl-4-butylbenzoat
Methyl-4-butylbenzoat
Formel:
C₁₂H₁₆O₂ |
MDL:
MFCD00277209 CAS-Nummer: 20651-69-8 EINECS: 243-939-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...