3-Isoxazolcarbonsäure ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Isoxazole-3-carboxylic Acid
H58438.03
H58438.06
H58438.03EA
145
EUR
H58438.03
H58438.06
3-Isoxazolcarbonsäure ≥97%
3-Isoxazolecarboxylic acid
Purity: 97%
CAS No.: 3209-71-0
Molecular Formula: C4H3NO3
Formula Weight: 113.07
Beilstein Reference: 110919
MDL No.: MFCD06738796
Formel:
C₄H₃NO₃ Molecular Weight: 113,07 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06738796 CAS-Nummer: 3209-71-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...