3-Methyl-4-isoxazolcarbonsäure

Lieferant: CHEMPUR

56-0019.1 56-0019.5
CHMP56-0019.1EA 0 EUR
CHMP56-0019.1 CHMP56-0019.5
3-Methyl-4-isoxazolcarbonsäure
3-Methyl-4-isoxazolecarboxylic acid
Formel: C₅H₅NO₃
MDL: MFCD01318161
CAS-Nummer: 17153-20-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR