5-Phenyl-3-isoxazolcarbonsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Phenylisoxazole-3-carboxylic acid

M-Clarity™ Qualitätsstufe = MQ100

645087-1G 645087-5G
SIAL645087-1GEA 51.4 EUR
SIAL645087-1G SIAL645087-5G
5-Phenyl-3-isoxazolcarbonsäure, Sigma-Aldrich®
5-Phenyl-3-isoxazolecarboxylic acid
Formel: C₁₀H₇NO₃
Molecular Weight: 189,1700 g/mol
MDL: MFCD00464221
CAS-Nummer: 14441-90-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR