2-Ethoxyisonicotinsäure
Lieferant: COMBI-BLOCKS
Synonyme:
2-Ethoxypyridine-4-carboxylic acid
CA-4046-5G
CA-4046-25G
CA-4046-1G
COBBCA-4046-5GEA
303
EUR
COBBCA-4046-5G
COBBCA-4046-25G
COBBCA-4046-1G
2-Ethoxyisonicotinsäure
2-Ethoxyisonicotinsäure
Formel:
C₈H₉NO₃ Molecular Weight: 167,1620 g/mol |
MDL:
MFCD04115719 CAS-Nummer: 91940-86-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...