2-Methoxyisonicotinsäure
Lieferant: COMBI-BLOCKS
Synonyme:
2-Methoxy-4-pyridinecarboxylic acid
CA-4045-1G
CA-4045-100G
CA-4045-5G
CA-4045-25G
COBBCA-4045-1GEA
21.6
EUR
COBBCA-4045-1G
COBBCA-4045-100G
COBBCA-4045-5G
COBBCA-4045-25G
2-Methoxyisonicotinsäure
2-Methoxyisonicotinsäure
Formel:
C₇H₇NO₃ Molecular Weight: 153,1400 g/mol |
MDL:
MFCD04115718 CAS-Nummer: 105596-63-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...