5-Amino-2-brombenzonitril
Lieferant: COMBI-BLOCKS
AN-1166-5G
AN-1166-1G
COBBAN-1166-5GEA
89.6
EUR
COBBAN-1166-5G
COBBAN-1166-1G
5-Amino-2-brombenzonitril
5-Amino-2-brombenzonitril
Formel:
C₇H₅BrN₂ Molecular Weight: 197,0320 g/mol |
MDL:
MFCD00234259 CAS-Nummer: 72115-09-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...