2-Fluor-6-(4-methylphenoxy)benzonitril
Lieferant: FLUOROCHEM
008345-1G
008345-5G
FLUO008345-1GEA
91.2
EUR
FLUO008345-1G
FLUO008345-5G
2-Fluor-6-(4-methylphenoxy)benzonitril
2-Fluor-6-(4-methylphenoxy)benzonitril
Formel:
C₁₄H₁₀FNO |
MDL:
MFCD00068203 CAS-Nummer: 175204-08-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...