2,6-Difluor-3-pyridinboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
2,6-Difluoropyridine-3-boronic acid
BB-2275-5G
BB-2275-1G
COBBBB-2275-5GEA
182
EUR
COBBBB-2275-5G
COBBBB-2275-1G
2,6-Difluor-3-pyridinboronsäure
2,6-Difluoro-3-pyridineboronic acid
Formel:
C₅H₄BF₂NO₂ Molecular Weight: 158.90 g/mol |
MDL:
MFCD03788245 CAS-Nummer: 136466-94-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...