3-Fluor-4-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3-Fluoropyridin-4-yl)boronic acid

BB-3440-1G BB-3440-5G
COBBBB-3440-1GEA 165 EUR
COBBBB-3440-1G COBBBB-3440-5G
3-Fluor-4-pyridinboronsäure
3-Fluoro-4-pyridineboronic acid
Formel: C₅H₅BFNO₂
Molecular Weight: 140.91 g/mol
MDL: MFCD03788558
CAS-Nummer: 458532-97-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR