3-Fluor-4-pyridinboronsäure Hydrat

Lieferant: COMBI-BLOCKS

BB-8435-5G BB-8435-1G
COBBBB-8435-5GEA 595 EUR
COBBBB-8435-5G COBBBB-8435-1G
3-Fluor-4-pyridinboronsäure Hydrat
3-Fluor-4-pyridinboronsäure Hydrat
Formel: C₅H₅BFNO₂·H₂O
MDL: MFCD03701686
CAS-Nummer: 1029880-18-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR