5-Fluor-3-pyridinboronsäure ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
5-Fluoropyridine-3-boronic acid, (5-Fluoropyridin-3-yl)boronic acid
H52860.MD
H52860.03
H52860.06
H52860.MDEA
74.1
EUR
H52860.MD
H52860.03
H52860.06
5-Fluor-3-pyridinboronsäure ≥98%
5-Fluoro-3-pyridineboronic acid
Purity: 98%
CAS No.: 872041-86-6
Molecular Formula: C5H5BFNO2
Formula Weight: 140.91
Melting Point: 233-236°
MDL No.: MFCD07368243
Formel:
C₅H₅BFNO₂ Molecular Weight: 140.91 g/mol Schmelzpunkt: 233…236 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07368243 CAS-Nummer: 872041-86-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...