5-Fluor-3-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 5-Fluoropyridine-3-boronic acid, (5-Fluoropyridin-3-yl)boronic acid

BB-3512-1G BB-3512-5G
COBBBB-3512-1GEA 54.8 EUR
COBBBB-3512-1G COBBBB-3512-5G
5-Fluor-3-pyridinboronsäure
5-Fluoro-3-pyridineboronic acid
Formel: C₅H₅BFNO₂
Molecular Weight: 140.91 g/mol
MDL: MFCD07368243
CAS-Nummer: 872041-86-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR