N,N-Diethyl-2-methylbenzamid

Lieferant: COMBI-BLOCKS

CA-4988-25G CA-4988-5G CA-4988-1G
COBBCA-4988-25GEA 1700 EUR
COBBCA-4988-25G COBBCA-4988-5G COBBCA-4988-1G
N,N-Diethyl-2-methylbenzamid
N,N-Diethyl-2-methylbenzamid
Formel: C₁₂H₁₇NO
MDL: MFCD00463019
CAS-Nummer: 2728-04-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR