2-Methyl-N-tert-butylbenzamid
Lieferant: COMBI-BLOCKS
Synonyme:
N-t-butyl-2-methylbenzamide
CA-4779-25G
CA-4779-5G
COBBCA-4779-25GEA
375
EUR
COBBCA-4779-25G
COBBCA-4779-5G
2-Methyl-N-tert-butylbenzamid
2-Methyl-N-tert-butylbenzamid
Formel:
C₁₂H₁₇NO |
MDL:
MFCD00463017 CAS-Nummer: 104847-07-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...