3-Methyl-N-tert-butylbenzamid

Lieferant: COMBI-BLOCKS

Synonyme: N-tert-Butyl-3-methylbenzamide

CA-4990-5G CA-4990-25G
COBBCA-4990-5GEA 149 EUR
COBBCA-4990-5G COBBCA-4990-25G
3-Methyl-N-tert-butylbenzamid
3-Methyl-N-tert-butylbenzamid
Formel: C₁₂H₁₇NO
MDL: MFCD00996285
CAS-Nummer: 42498-33-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR