4-Methyl-N-tert-butylbenzamid

Lieferant: COMBI-BLOCKS

Synonyme: N-tert-Butyl-4-methylbenzamide

CA-4776-25G CA-4776-5G
COBBCA-4776-25GEA 375 EUR
COBBCA-4776-25G COBBCA-4776-5G
4-Methyl-N-tert-butylbenzamid
4-Methyl-N-tert-butylbenzamid
Formel: C₁₂H₁₇NO
MDL: MFCD00463097
CAS-Nummer: 42498-32-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR