4-Brom-8-methylchinolin

Lieferant: COMBI-BLOCKS

HC-2763-5G HC-2763-1G HC-2763-250M
COBBHC-2763-5GEA 912 EUR
COBBHC-2763-5G COBBHC-2763-1G COBBHC-2763-250M
4-Brom-8-methylchinolin
4-Bromo-8-methylquinoline
Formel: C₁₀H₈BrN
Molecular Weight: 222,0800 g/mol
MDL: MFCD08063210
CAS-Nummer: 36075-68-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR