7-Bromo-8-methylquinoline

Lieferant: COMBI-BLOCKS

Synonyme: 7-Bromo-8-methyl-1-azanaphthalene

HC-2810-5G HC-2810-1G
COBBHC-2810-5GEA 358 EUR
COBBHC-2810-5G COBBHC-2810-1G
7-Bromo-8-methylquinoline
7-Bromo-8-methylquinoline
Formel: C₁₀H₈BrN
Molecular Weight: 222,081 g/mol
MDL: MFCD09907849
CAS-Nummer: 809248-61-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR