3-Chlor-6-phenylpyridazin

Lieferant: Thermo Scientific

Synonyme: 3-Phenyl-6-chloropyridazine

BTB12369.1 BTB12369.10
MAYBBTB12369.1EA 0 EUR
MAYBBTB12369.1 MAYBBTB12369.10
3-Chlor-6-phenylpyridazin
3-Chloro-6-phenylpyridazine
Formel: C₁₀H₇ClN₂
Molecular Weight: 190,6300 g/mol
MDL: MFCD00075253
CAS-Nummer: 20375-65-9
EINECS: 243-772-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR