Ethyltrifluorpyruvat

Lieferant: FLUOROCHEM

Synonyme: Ethyl 3,3,3-trifluoro-2-oxopropanoate, Ethyl 3,3,3-trifluoropyruvate

003224-25G 003224-10G
FLUO003224-25GEA 97.1 EUR
FLUO003224-25G FLUO003224-10G
Ethyltrifluorpyruvat
Ethyltrifluorpyruvat
Formel: C₅H₅F₃O₃
Molecular Weight: 170.09 g/mol
MDL: MFCD00114935
CAS-Nummer: 13081-18-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR