5,6-Dichlor-2-methylbenzimidazol

Lieferant: COMBI-BLOCKS

HC-3145-5G HC-3145-25G HC-3145-1G
COBBHC-3145-5GEA 279 EUR
COBBHC-3145-5G COBBHC-3145-25G COBBHC-3145-1G
5,6-Dichlor-2-methylbenzimidazol
5,6-Dichloro-2-methylbenzimidazole
Formel: C₈H₆Cl₂N₂
Molecular Weight: 201,0500 g/mol
MDL: MFCD00276973
CAS-Nummer: 6478-79-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR