6-Fluor-1-methylbenzimidazol

Lieferant: COMBI-BLOCKS

Synonyme: 6-Fluoro-1-methyl-1H-benzo[d]imidazole

IN-0185-5G IN-0185-1G
COBBIN-0185-5GEA 1040 EUR
COBBIN-0185-5G COBBIN-0185-1G
6-Fluor-1-methylbenzimidazol
6-Fluoro-1-methylbenzimidazole
Formel: C₈H₇FN₂
Molecular Weight: 150.15 g/mol
MDL: MFCD12546503
CAS-Nummer: 1187385-86-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR