5-Brom-2-hydroxybenzonitril, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

697184-1G 697184-5G
SIAL697184-1GEA 32.9 EUR
SIAL697184-1G SIAL697184-5G
5-Brom-2-hydroxybenzonitril, Sigma-Aldrich®
5-Brom-2-hydroxybenzonitril
Formel: C₇H₄BrNO
Molecular Weight: 198,02 g/mol
MDL: MFCD00143428
CAS-Nummer: 40530-18-5
EINECS: 254-958-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR