4-Brom-2-hydroxybenzonitril

Lieferant: COMBI-BLOCKS

OS-7573-5G OS-7573-25G OS-7573-1G
COBBOS-7573-5GEA 0 EUR
COBBOS-7573-5G COBBOS-7573-25G COBBOS-7573-1G
4-Brom-2-hydroxybenzonitril
4-Brom-2-hydroxybenzonitril
Formel: C₇H₄BrNO
Molecular Weight: 198.01 g/mol
MDL: MFCD07780675
CAS-Nummer: 288067-35-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR