2,4-Dibrom-6-hydroxybenzonitril

Lieferant: CHEMPUR

FM00045.250 FM00045.1
CHMPFM00045.250EA 0 EUR
CHMPFM00045.250 MAYBFM00045.1
2,4-Dibrom-6-hydroxybenzonitril
2,4-Dibrom-6-hydroxybenzonitril
Formel: C₇H₃Br₂NO
CAS-Nummer: 73289-86-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR