3,5-Dibrom-2-hydroxybenzonitril

Lieferant: CHEMPUR

RH00917.250 RH00917.1
CHMPRH00917.250EA 0 EUR
CHMPRH00917.250 MAYBRH00917.1
3,5-Dibrom-2-hydroxybenzonitril
3,5-Dibrom-2-hydroxybenzonitril
Formel: C₇H₃Br₂NO
Molecular Weight: 276.9128 g/mol
MDL: MFCD00662354
CAS-Nummer: 40718-08-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR