tert-Butyl-2-chlor-4-fluorbenzoat

Lieferant: COMBI-BLOCKS

Synonyme: tert-Butyl 2-chloro-4-fluorobenzoate

CA-4288-1G CA-4288-5G CA-4288-25G
COBBCA-4288-1GEA 149 EUR
COBBCA-4288-1G COBBCA-4288-5G COBBCA-4288-25G
tert-Butyl-2-chlor-4-fluorbenzoat
tert-Butyl-2-chlor-4-fluorbenzoat
Formel: C₁₁H₁₂ClFO₂
Molecular Weight: 230.6632 g/mol
MDL: MFCD09800898
CAS-Nummer: 911314-43-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR