tert-Butyl-4-chlor-2-fluorbenzoat

Lieferant: COMBI-BLOCKS

CA-4290-5G CA-4290-25G CA-4290-1G
COBBCA-4290-5GEA 451 EUR
COBBCA-4290-5G COBBCA-4290-25G COBBCA-4290-1G
tert-Butyl-4-chlor-2-fluorbenzoat
tert-Butyl-4-chlor-2-fluorbenzoat
Formel: C₁₁H₁₂ClFO₂
MDL: MFCD09475908
CAS-Nummer: 941294-14-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR