5-Chlor-2,4-difluorbenzolsulfonamid

Lieferant: FLUOROCHEM

CFF906. 005525-1G 005525-2.5G
CHMPCFF906.EA 0 EUR
CHMPCFF906. FLUO005525-1G FLUO005525-2.5G
5-Chlor-2,4-difluorbenzolsulfonamid
5-Chlor-2,4-difluorbenzolsulfonamid
Formel: C₆H₄ClF₂NO₂S
MDL: MFCD01940427
CAS-Nummer: 13656-56-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR