4-Chlor-2,5-difluorbenzolsulfonamid

Lieferant: FLUOROCHEM

CFF905. 005524-1G 005524-2.5G
CHMPCFF905.EA 0 EUR
CHMPCFF905. FLUO005524-1G FLUO005524-2.5G
4-Chlor-2,5-difluorbenzolsulfonamid
4-Chlor-2,5-difluorbenzolsulfonamid
Formel: C₆H₄ClF₂NO₂S
MDL: MFCD02091387
CAS-Nummer: 287172-62-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR