N,N-Diethyl 4-fluorbenzolsulfonamid

Lieferant: COMBI-BLOCKS

OT-1117-5G OT-1117-1G
COBBOT-1117-5GEA 0 EUR
COBBOT-1117-5G COBBOT-1117-1G
N,N-Diethyl 4-fluorbenzolsulfonamid
N,N-Diethyl 4-fluorbenzolsulfonamid
Formel: C₁₀H₁₄FNO₂S
MDL: MFCD01213080
CAS-Nummer: 309-91-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR