4-Brom-N,N-diethyl-3-(trifluormethyl)benzolsulfonamid

Lieferant: COMBI-BLOCKS

HC-6470-5G HC-6470-1G HC-6470-250M
COBBHC-6470-5GEA 2430 EUR
COBBHC-6470-5G COBBHC-6470-1G COBBHC-6470-250M
4-Brom-N,N-diethyl-3-(trifluormethyl)benzolsulfonamid
4-Brom-N,N-diethyl-3-(trifluormethyl)benzolsulfonamid
Formel: C₁₁H₁₃BrF₃NO₂S
MDL: MFCD09972159
CAS-Nummer: 1020253-03-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR