4-Brom-N-ethyl-3-(trifluormethyl)benzolsulfonamid

Lieferant: COMBI-BLOCKS

HC-6465-5G HC-6465-1G
COBBHC-6465-5GEA 983 EUR
COBBHC-6465-5G COBBHC-6465-1G
4-Brom-N-ethyl-3-(trifluormethyl)benzolsulfonamid
4-Brom-N-ethyl-3-(trifluormethyl)benzolsulfonamid
Formel: C₉H₉BrF₃NO₂S
MDL: MFCD09972155
CAS-Nummer: 1020252-99-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR