4-Brom-N-methyl-3-(trifluormethyl)benzolsulfonamid

Lieferant: COMBI-BLOCKS

HC-6467-1G HC-6467-5G
COBBHC-6467-1GEA 404 EUR
COBBHC-6467-1G COBBHC-6467-5G
4-Brom-N-methyl-3-(trifluormethyl)benzolsulfonamid
4-Brom-N-methyl-3-(trifluormethyl)benzolsulfonamid
Formel: C₈H₇BrF₃NO₂S
MDL: MFCD09972157
CAS-Nummer: 1020253-01-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR