3'-Chlor-2'-fluor-5'-(trifluormethyl)acetophenon

Lieferant: CHEMPUR

FL5673.1 FL5673.5
CHMPFL5673.1EA 0 EUR
CHMPFL5673.1 CHMPFL5673.5
3'-Chlor-2'-fluor-5'-(trifluormethyl)acetophenon
3'-Chlor-2'-fluor-5'-(trifluormethyl)acetophenon
Formel: C₉H₅ClF₄O
MDL: MFCD01631398
CAS-Nummer: 261763-00-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR