tert-Butyl-2-brom-5-methoxybenzoat

Lieferant: COMBI-BLOCKS

CA-4295-1G CA-4295-5G CA-4295-25G
COBBCA-4295-1GEA 149 EUR
COBBCA-4295-1G COBBCA-4295-5G COBBCA-4295-25G
tert-Butyl-2-brom-5-methoxybenzoat
tert-Butyl-2-brom-5-methoxybenzoat
Formel: C₁₂H₁₅BrO₃
MDL: MFCD09878358
CAS-Nummer: 957063-12-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR