Methyl-3-brom-5-ethoxybenzoat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 3-bromo-5-ethoxybenzoate

CA-4982-5G CA-4982-1G
COBBCA-4982-5GEA 303 EUR
COBBCA-4982-5G COBBCA-4982-1G
Methyl-3-brom-5-ethoxybenzoat
Methyl-3-brom-5-ethoxybenzoat
Formel: C₁₀H₁₁BrO₃
MDL: MFCD16295085
CAS-Nummer: 860695-64-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR