1-Brom-3,5-difluor-4-iodbenzol

Lieferant: COMBI-BLOCKS

Synonyme: 5-Bromo-1,3-difluoro-2-iodobenzene

OS-7662-5G OS-7662-25G OS-7662-1G
COBBOS-7662-5GEA 50.8 EUR
COBBOS-7662-5G COBBOS-7662-25G COBBOS-7662-1G
1-Brom-3,5-difluor-4-iodbenzol
1-Bromo-3,5-difluoro-4-iodobenzene
Formel: C₆H₂BrF₂I
Molecular Weight: 318,8854 g/mol
MDL: MFCD01861126
CAS-Nummer: 160976-02-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR