2,4,5-Trifluoriodbenzol
Lieferant: FLUOROCHEM
Synonyme:
2,4,5-Trifluoro-1-iodobenzene, 1,2,4-Trifluoro-5-iodobenzene
021383-1G
021383-5G
FLUO021383-1GEA
31.1
EUR
FLUO021383-1G
FLUO021383-5G
2,4,5-Trifluoriodbenzol
2,4,5-Trifluoriodbenzol
Formel:
C₆H₂F₃I Molecular Weight: 257.98 g/mol |
MDL:
MFCD04973898 CAS-Nummer: 17533-08-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...